Order Entry
ContactUsLinkComponent
VWR* Disposable Serological Pipets
VWR® Disposable Serological Pipettes, Glass
Catalog #: 93000-686
Supplier:  VWR International
VWR* Disposable Serological Pipets
VWR® Disposable Serological Pipettes, Glass
Catalog #: 93000-686
Supplier:  VWR International

Specifications

  • Capacity:
    5 ml
  • Packaging:
    Canister Pack, Plugged, Sterile
  • Tolerance:
    ±0.15 ml
  • Subdivisions:
    0.1 mL
  • Negative Graduations:
    1 ml

Specifications

About this item

VWR® Disposable Serological Pipettes are available in sterile or nonsterile formats, with a variety of packaging options to choose from.

  • Uniform pulled tip, calibrated to deliver
  • Borosilicate glass construction
  • Negative graduations increase versatility
  • Available in sterile or nonsterile formats

These pipettes have a uniform pulled tip and are calibrated to deliver to the tip, with two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered. Negative graduations increase the versatility of the pipet. Pipettes are available sterile and plugged or in bulk packs of nonsterile, non-plugged pipettes. Nonsterile, non-plugged pipettes feature a blue scale. Made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. Individually wrapped, multi-pack, and canister pack pipettes are also designed from ASTM Specification E714.

Packaging: Individually wrapped pipettes are packaged in color-coded paper/plastic wraps and overpacked in convenient dispensing shelf packs for laboratory benches. Multi-pack pipettes come in color-coded bags of 25 pipettes (1 ml and 2 ml sizes) or 10 pipettes (5 ml and 10 ml sizes). Canister pack pipettes are packaged in a reclosable pack of 50 pieces each sealed in a plastic bag. Bulk pack pipettes are sealed in plastic bags and overpacked in shelf packs.
Customers Who Bought This Also Bought