CAP 11MM SNAP DK BLU CUT T/S
Catalog #: 89200-106
Supplier: DWK Life Sciences (KIMBLE)
Customers Who Bought This Also Bought
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$599.10 to $2,396.15