Disposable Automation Reservoirs
Catalog #: 89200-086
Supplier: Thermo Fisher Scientific
Specifications
- Description:384 channel
- Supplier No.:1064-05-7
Specifications
About this item
Disposable automation reservoirs are of polypropylene construction and are designed for simple, economical handling of reagents on automated systems.
The standard microplate dimensions enables reservoirs to be used in plate handlers or stackers.
With the same dimensions as standard microplates, these disposable reservoirs can be used in plate handlers or stackers. The removable splash baffles prevent reagents from splashing out when being moved on robotic decks. They are compatible with virtually any automated liquid handler as well as all of our automated pipette tips.
Customers Who Bought This Also Bought