ContactUsLinkComponent
Brand* Handystep* S Repeating Pipette, Brandtech
BRAND HandyStep® S Repeating Pipette, BrandTech
Catalog #: 89176-652
Supplier:  Brandtech
Brand* Handystep* S Repeating Pipette, Brandtech
BRAND HandyStep® S Repeating Pipette, BrandTech
Catalog #: 89176-652
Supplier:  Brandtech
Specifications
  • Description:
    BRAND HandyStep® Repeating Pipette
  • Cat. no.:
    89176-652
  • Supplier No.:
    705110
Specifications
About this item
The BRAND HandyStep® S repeating pipette provides unsurpassed versatility in a purely mechanical repeating pipette.
Comfortable. The slim design of the HandyStep® S is lightweight, well-balanced, and provides ambidextrous one-handed volume selection and dispensing. A new Tip ejector provides contact-free tip removal.
Innovative polymer construction provides a dependable, corrosion resistant, robust instrument. The purely mechanical design means no batteries to charge or replace–a convenient integrated volume table provides a quick reference for dispensed volumes.
Included shelf/rack mount also attaches easily to BRAND Transferpette® S carousel stand.
In conjunction with BRAND PD-Tip™ syringe tips, the rugged HandyStep S offers nearly sixty different volume settings with accuracy and precision.
Ordering information: The HandyStep® S is supplied in recyclable packaging with performance certificate, shelf/rack mount, one each 0.1mL, 1mL and 10mL tips, operating manual and two year warranty.
Customers Who Bought This Also Bought