Dissolution Calibration Tools, Quality Lab Accessories
Catalog #: 89173-172
Supplier: QLA
Specifications
- Description:Digital Verticality Meter, Multi-Instrument
- Cat. no.:89173-172
- Supplier No.:VERTMET-MI
Specifications
About this item
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
- Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
Certifications: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
Customers Who Bought This Also Bought