Order Entry
ContactUsLinkComponent
SPECIMEN TRANSPORT BAG 100 PK
Specimen Transport Bag with Requistion Pouch, Therapak®
Catalog #: 89171-048
Supplier:  Therapak, LLC
SPECIMEN TRANSPORT BAG 100 PK
Specimen Transport Bag with Requistion Pouch, Therapak®
Catalog #: 89171-048
Supplier:  Therapak, LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Transport bag with biohazard symbol and absorbent
  • Cat. no.:
    89171-048
  • Supplier No.:
    40007

Specifications

About this item

These 6×9" polyethylene specimen bags feature a removable, adhesive release liner that secures specimen containers in a liquid-tight primary pouch.

  • Fold and insert requisition in second pouch.

Once the bag is sealed, both pouches are tamper-evident which provides protection of specimen and sensitive patient information during the transport process.

Delivery: Each bag comes with a 3×4" absorbent sheet.
Customers Who Bought This Also Bought