ContactUsLinkComponent
VWR Carboy Cap
Accessories for VWR® Carboys with Versatile Caps
Catalog #: 89170-834
Supplier:  VWR International
VWR Carboy Cap
Accessories for VWR® Carboys with Versatile Caps
Catalog #: 89170-834
Supplier:  VWR International
Specifications
  • Description:
    Open cap with adapter, 80 mm, with two ¹/₂" hose barbs and vent
  • Cat. no.:
    89170-834