ContactUsLinkComponent
Loose Fitting Facepiece, Tychem Qc, Large
Face Pieces, Loose-Fitting, Bullard
Catalog # 89168-708
Supplier:  Bullard
Face Pieces, Loose-Fitting, Bullard
Catalog # 89168-708
Supplier:  Bullard
Supplier Number:  20LF2L
Specifications
  • Description:
    Loose-Fitting Facepiece with Narrow Profile, Large
  • Size:
    71/2 81/2"
  • Cat. no.:
    89168-708
  • Supplier No.:
    20LF2L
Specifications
About this item
The latex-free facepiece provides comfortable, affordable, and effective protection from a wide range of respiratory hazards. Available for use as a CC20 Series supplied air or EVA Series powered air-purifying respirator, this loose fitting facepiece constructed of DuPont™ Tychem® QC provides its users with versatility, advanced features, and ease of use.
  • Soft Plenum™ Delivery System delivers air to the worker’s breathing zone; provides more airflow where it is most needed and helps to prevent fogging
  • Sewn-in soft suspension provides a comfortable fit to the worker’s head while allowing for complete disposability of the facepiece
  • Twist-lock breathing tube connection provides ease of use by allowing quick connection or disassembly of the breathing tube
  • Wide Angle Lens gives the worker a wide panoramic field of vision eliminating the“closed-in” feeling of many respirators
  • Climate Control Option When used as a CC20 Series airline respirator, the Loose Fitting Facepiece may be used with the optional climate control devices.
The Loose Fitting Facepiece is used in a wide variety of applications including pharmaceutical manufacturing, spray foam, general industry, painting, healthcare , woodworking.
Certifications: NIOSH approved
Packaging: 10 face pieces per case.
Frequently Bought Together