Glass Cell Spreaders, Chemglass
Catalog #: 89166-788
Supplier: Chemglass
Specifications
- Cat. no.:89166-788
- Style:90° Bend
- Supplier No.:CLS-1350-02
Specifications
About this item
These glass cell spreaders are made of 5 mm borosilicate glass
Spreaders are ideal for spreading inoculums over the agar face in Petri dish cultures.
Customers Who Bought This Also Bought