Suba·Seal® Septa, Ace Glass
Catalog #: 89166-144
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Suba·Seal Septa®
- Material:Red Rubber
- Size:#45, Fits 24/40 and 24/25 standard taper joints
- Cat. no.:89166-144
- Fits Neck Size:22 mm I.D
- Supplier No.:9107-142
Specifications
About this item
The Suba·Seal® Septa is a high quality rubber septa designed with serrated bottom for sealing open-top glass vessels as well as standard taper joints
Suba·Seal® Septa have serrated sealing capability to the inside of the vessel, as well as the turn-over top seal. Autoclaveable and packaged in white-room conditions.
Ideal for moisture or air sensitive applications.
Ordering information: Suba-Seal® is a trademark of Sigma-Aldrich Biotechnology LP.
Customers Who Bought This Also Bought