Order Entry
ContactUsLinkComponent
Cell-Id* Autophagy Detection Kit 1 Kit
Cyto-ID® Autophagy Detection Kit, Enzo Life Sciences
Catalog #: 89165-926
Supplier:  Enzo Life Sciences
CAS Number:  
Cell-Id* Autophagy Detection Kit 1 Kit
Cyto-ID® Autophagy Detection Kit, Enzo Life Sciences
Catalog #: 89165-926
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cell-ID® Autophagy Detection Kit
  • Size:
    1 Kit
  • Storage conditions:
    −80 °C
  • Application:
    Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection and HTS
  • Cat. no.:
    89165-926
  • Supplier No.:
    51031-K200

Specifications

About this item

The Cyto-ID® Autophagy detection kit provides a rapid, specific and quantitative approach for monitoring autophagy in live cells by fluorescence microscopy and flow cytometry.

Cyto-ID® Autophagy Detection Kit measures autophagic vacuoles and monitors autophagic flux in live cells using a novel dye that selectively labels autophagic vacuoles. The dye has been optimized through the identification of titratable functional moieties that allow for minimal staining of lysosomes while exhibiting bright fluorescence upon incorporation into pre-autophagosomes, autophagosomes, and autolysosomes (autophagolysosomes).

Delivery: Kit contains Cyto-ID® Green Detection Reagent, 50 µL, Hoechst 33342 Nuclear Stain, 50 µL Autophagy Inducer (Rapamycin, 25 nmol) and 10X Assay Buffer, 30 mL.
Customers Who Bought This Also Bought