Order Entry
ContactUsLinkComponent
Omnimmp Fluorescent Substrate 1 Mg
OMNIMMP® Fluorescent Substrate
Catalog #: 89160-966
Supplier:  Enzo Life Sciences
CAS Number:  
Omnimmp Fluorescent Substrate 1 Mg
OMNIMMP® Fluorescent Substrate
Catalog #: 89160-966
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 mg
  • Protein/peptide name:
    Fluorescent Substrate
  • Purity:
    98%
  • Cat. no.:
    89160-966
  • Supplier No.:
    P126-0001

Specifications

About this item

Highly quenched and sensitive fluorogenic MMP substrate.

Fluorogenic substrate for nearly all matrix metalloproteinase (MMP) enzymes, and ADAM17/TACE. This highly-quenched (99.5%) substrate is useful for inhibitor screening and kinetic analysis. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have been used: 320 – 340/400 – 420. Also available: OmniMMPTM Fluorogenic Control Peptide (BML-P127).

Customers Who Bought This Also Bought