L(+)-Arginine ≥98% (by TLC)
Catalog #: 89142-488
Supplier: Enzo Life Sciences
CAS Number: 74-79-3
Specifications
- Size:25 g
- Cat. no.:89142-488
- Supplier No.:101-004-G025
Specifications
Specification Test Results
- L(+)-Arginine ≥98% (by TLC):
Specification Test Results
About this item
Synonyms: H-L-Arg-OH,(S)-2-Amino-5-guanidinopentanoic acid,Arginine,(S)-(+)-Arginine,L-Arginine
:L(+)-Arginine
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 238 °C
:Flash Pt:
:Flash Pt:
:M.W. 174.2 g/mol
:238
:368
Customers Who Bought This Also Bought