To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Reactivity:Human
- Sensitivity:50 pg/ml (range 62.5 - 4,000 pg/ml)
- Cat. no.:89141-870
- Supplier No.:EKS-895
- No. of Tests:96 Wells
Specifications
About this item
Most sensitive, rapid ELISA kit for highly specific, quantitative detection of human HSP90α.
For the quantitative determination of HSP90α in cell lysates, serum, and tissue of human origin.
The HSP90α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.