Order Entry
ContactUsLinkComponent
Protein A EIA Kit 1 X 96 Wells
Protein A ELISA Kit, Enzo Life Sciences
Catalog #: 89141-106
Supplier:  Enzo Life Sciences
CAS Number:  
Protein A EIA Kit 1 X 96 Wells
Protein A ELISA Kit, Enzo Life Sciences
Catalog #: 89141-106
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Sensitivity:
    9.01pg/mL (range 15.63 - 1,000pg/mL)
  • Cat. no.:
    89141-106
  • Supplier No.:
    900-057
  • No. of Tests:
    96 Wells

Specifications

About this item

Most sensitive (9.01pg/ml) colorimetric ELISA for monitoring residual contamination.

For the quantitative determination of residual protein A contamination in protein A purified IgG preparations from any species.

The Protein A EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ultra-sensitive quantification (< 1 ppm) of Protein A residuals in purified humanized monoclonal antibody preparations. This kit recognizes four different Protein A constructs.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate,Standard, TMB Substrate, Stop solution 2.
Customers Who Bought This Also Bought