VWR® Safety-Vented™ Wide Mouth Labeled Wash Bottles, LDPE
Catalog #: 89141-048
Supplier: VWR International
Specifications
- Capacity:500 ml (16.9 oz.)
- Description:Ethanol
- Cat. no.:89141-048
- Cap Color:Natural
Specifications
About this item
Wide mouth, color-coded, low-density polyethylene wash bottles feature a unique two-piece polypropylene venting assembly that prevents pressure buildup of solvents inside the bottle.
- Pre-labeled and vented for maximum safety
- Eliminate solvent drips
- Easy-to-read labeling
Venting assembly eliminates solvent drips from the spout due to internal evaporation. Easy-to-read solvent labeling includes DOT, NFPA codes and symbols, as well as CAS numbers. Bottles are leakproof.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Caution: LDPE wash bottles are intended for short-term dispensing use only and not for long-term storage. Bottles marked LYOB (Label Your Own Bottle) are designed for use only with solvents compatible with LDPE.
Customers Who Bought This Also Bought