ContactUsLinkComponent
VWR Economy Timer
VWR® Economy Timer
Catalog #: 89140-544
Supplier:  VWR International
VWR® Economy Timer
Catalog #: 89140-544
Supplier:  VWR International
Specifications
  • Description:
    Economy timer
  • Cat. no.:
    89140-544
Specifications
About this item
Timer can be programmed in countdown mode or countup mode up to 99 minutes, 59 seconds.
  • Count up and down simultaneously
  • Big digit display
Delivery: Supplied with loud buzzer, magnet, clip, and stand.
Customers Who Bought This Also Bought