96-Well Plates, Polypropylene, With Lids, Greiner Bio-One
Catalog #: 89140-484
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Color:Clear
- Material:PP
- Well Shape:Round-Bottom
- Supplier No.:650261-656
Specifications
About this item
96 Well Plates feature polypropylene round (U) bottom, chimney style, clear with condensation ring lid to meet your assay and storage needs.
- Condensation ring design
- Chimney-style molding
- Round bottom
These lids are suitable for use with live cells, and are free of detectable DNase, RNase, human DNA, and pyrogens.
Ordering information: Paltes are packeged 8 per pack. Sterile Case of 80
Customers Who Bought This Also Bought