ContactUsLinkComponent
CF770 succinimidyl ester (CF770 SE)
Reactive CF® Dyes and Protein Labeling Kits, Biotium
Catalog #: 89139-842
Supplier:  Biotium
CAS Number:  
Reactive CF® Dyes and Protein Labeling Kits, Biotium
Catalog #: 89139-842
Supplier:  Biotium
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CF®770 succinimidyl ester
  • Size:
    1 µmol
  • Cat. no.:
    89139-842
  • Supplier No.:
    92150
Specifications
About this item
Biotium offers a number of reactive dye formats for labelling proteins, nucleic acids, and other biomolecules with fluorescent dyes. CF® dyes offer advantages in brightness and photostability compared other similar commercial dyes. CF® dyes are available in a number of reactive dye formats for CF® dye colours span the visible and near-infrared spectra.
Amine groups react with activated carboxylic acids. Aminooxy (also known as hydroxylamine) groups readily react with aldehydes or ketones without the need for a reducing agent. Hydrazides can be microinjected into cells as fixable polar tracers for visualization of neuronal morphology. Hydrazides also can be used for labeling aldehyde or ketone groups, such as carbohydrate molecules after peroxidation with periodate. However, for this application we recommend using CF dye aminooxy forms, which are more reactive toward these groups than hydrazide forms. Maleimide reactive dyes label thiols, such as cysteine residues in proteins, and can be used as an alternative to SE labeling in cases where lysine modification alters the biological activity of the labeled protein. Succinimidyl ester (SE) groups label free amines, such as lysine residues in proteins, and are commonly used to label antibodies. CF® dye SE groups are more resistant to hydrolysis compared to most commercially available SE dyes, resulting in improved stability and labeling efficiency. SE Protein Labeling Kits include CF® dye SE and other components required to label and purify proteins. The kits are sufficient for three labeling reactions of 1 mg protein each.
CF® dye technologies are covered by pending US and international patents. CF® is a registred trademark of Biotium, Inc. AlexaFluor® is a registered trademark of Molecular Probes, Inc.
Biotium's line of CF® dyes offer advantages in brighness and/or photostability compared to Alexa Fluor® dyes and other similar commercial dyes. CF™ dyes are available in a number of reactive dye formats for CF™ dye colors span the visible and near-infrared spectra. See the CF® Dye Reference Table under the Datasheets tab to select the dye colors that are optimal for your application and instrument.
Ordering information: Succinimidyl Ester, Maleimide, and SE Protein Labeling Kits include a detailed protocol for labeling IgG or other proteins. See also: Mix-n-Stain™ Antibody Labeling Kits for rapid conjugation of 5 to 100 µg antibody.
Customers Who Bought This Also Bought