Order Entry
ContactUsLinkComponent
Fast Probe Master Mix(500rxn)
Fast Probe Master Mixes, Biotium
Catalog #: 89139-004
Supplier:  Biotium
CAS Number:  
Fast Probe Master Mix(500rxn)
Fast Probe Master Mixes, Biotium
Catalog #: 89139-004
Supplier:  Biotium
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    qPCR probe
  • Description:
    Fast Probe Master Mix
  • Size:
    500 reactions, 5 x 1 mL
  • Cat. no.:
    89139-004
  • Supplier No.:
    31005-1

Specifications

About this item

A qPCR master mix containing Cheetah™ Taq hotstart DNA polymerase, suitable for probe-based real-time PCR applications.

  • 2X probe-based qPCR master mix for fast cycling protocols
  • Suitable for all fluorescent probe types, including TaqMan®, BHQ®, and Molecular Beacon
  • Contains Cheetah™ Taq hotstart DNA polymerase

Fast Probe Master Mix is designed for fluorescent probe-based PCR applications. It is compatible with probes such as a TaqMan®, BHQ®, and Molecular Beacon. However for most PCR applications we recommend the Forget-Me-Not™ Universal Probe Master Mix, for superior activity.

Fast Probe Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.

Customers Who Bought This Also Bought