Order Entry
ContactUsLinkComponent
Coelenterazine cp
Coelenterazine Substrates for Renilla Luciferase and Apoaequorin, Biotium
Catalog #: 89138-306
Supplier:  Biotium
CAS Number:  
Coelenterazine Substrates for Renilla Luciferase and Apoaequorin, Biotium
Catalog #: 89138-306
Supplier:  Biotium
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Coelenterazine cp
  • Size:
    250 µG
  • Molecular Weight:
    415.5 g/mol
  • Supplier No.:
    10112-2

Specifications

About this item

Coelenterazine and its analogs are luminescent enzyme substrates for apoaequorin and Renilla luciferase.

Coelenterazine and its analogs are luminescent enzyme substrates for apoaequorin and Renilla luciferase. Apoaequorin is used as a calcium indicator whereas Renilla luciferase is commonly used as a reporter of transcription regulation. Coelenterazine binds to apoaequorin to form aequorin which emits light upon binding to calcium. Aequorin can be used to measure a broad concentration range of calcium from ~0.1 uM to >100 uM. Renilla luciferase has been widely used as a reporter protein and as a bioluminescence donor in bioluminescence resonance energy transfer (BRET) to study protein-protein interactions. Other uses of coelenterazine include chemiluminescent detection of superoxide anion and peroxynitrite in cells or tissues.

Can’t decide which coelenterazine analog to use? Try the Coelenterazine Sampler Kit (catalog no. 89138-348). The kit includes 25 ug each of nine coelenterazine analogs including native, cp, f, fcp, h, hcp, i, ip and n.

There are over a dozen of coelenterazine analogs that have been synthesized, many of which are now commercially available from Biotium. These coelenterazine analogs have different properties in terms of emission wavelength, cell membrane permeability and quantum efficiency.

Biotium offers coelenterazine 400a (also known as DeepBlue C™) (catalog no. 89138-350) which serves as a substrate for Renilla luciferase (Rluc) and generates an emission peak centered around 400 nm. It is the preferred Renilla luciferase substrate for BRET studies because it has minimal interference with the emission of the GFP acceptor.
Also, see Biotium's water soluble Aquaphile™ Coelenterazines, sold as coelenterazine native (catalog no. 89411-152) or coelenterzine h analog (catalog no. 89411-156). These formulations readily dissolve in water or buffer for in vivo use.
Lastly, Biotium offers methyl coelenterazine (coelenterazine, 2-methyl analog) (catalog no. 89138-344) which has been reported to be a superior antioxidant for cells against reactive oxygen species (ROS) such as singlet oxygen and superoxide anion. The coelenterazine derivative is membrane-permeant, non-toxic and highly reactive toward ROS.

Customers Who Bought This Also Bought