Order Entry
ContactUsLinkComponent
VACUSAFE Filter
Accessories for VACUSAFE™ Vacuum Aspiration System, INTEGRA Biosciences
Catalog #: 89136-838
Accessories for VACUSAFE™ Vacuum Aspiration System, INTEGRA Biosciences
Catalog #: 89136-838

Specifications

  • Description:
    VACUSAFE™ Hydrophobic Filter, 60mm, 0.45µm.
  • Cat. no.:
    89136-838
  • Supplier No.:
    158015

Specifications

About this item

Customers Who Bought This Also Bought