VWR® Sealing Film, Polyester
Catalog #: 89134-432
Supplier: VWR International
Specifications
- Optical property:Transparent
- Sterility:Sterile
- Description:Polyester sealing film
- Material:Polyester
- Thickness:49,5 µm
- Cat. no.:89134-432
Specifications
About this item
Polyester film, acrylic adhesive. These 49.5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
- Recommended temperature range from −40 to +120 °C
- Available sterile and non sterile
- Marginally pierceable by single channel pipette
- Applications: Short-term storage, incubation, ELISA
Each film L×D: 141,0×79,4 mm
Customers Who Bought This Also Bought