Order Entry
ContactUsLinkComponent
27G Blunt Needle 1.5In Sterile
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Catalog #: 89134-182
27G Blunt Needle 1.5In Sterile
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Catalog #: 89134-182
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Length:
    38 mm (1.5")
  • Sterility:
    Sterile
  • Cat. no.:
    89134-182
  • Packaging:
    Individually Wrapped
  • Supplier No.:
    B27-150

Specifications

About this item

These Luer Stub Adaptors (Blunt Needles) offer superior quality at an affordable price. With a plastic Luer hub and stainless-steel cannula, these blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters and dispensing of liquids. The smooth and burr-free stainless-steel needles will not damage the inner lumen when accessing catheters or tubing. Ensure the integrity of the catheter when dosing, sampling, flushing, and locking with these blunt needles.

  • Sterile needles are EtO exposed and individually packaged (25 per pack)
  • Non sterile needles are affordably packaged in bulk quantities of 100 or 1000
  • High-quality, low cost
  • Stainless steel cannula has a burr-free tip
  • Also available attached to empty or pre-filled flushing or locking syringes

Blunt Tip Needles (Luer Stub Adaptors) have countless laboratory applications. For instance, when dosing, sampling, or infusing, these blunt needles ensure a secure and reliable connection to in-dwelling catheters. The superior manufactured blunt needles provide a smooth, burr-free needle edge which prevents inadvertent damage to the lumen of the catheter caused by inferior needles. These high-quality, low-cost blunt needles are a critical tool for dosing, sampling, flushing, and locking catheters.

Ordering information: Contact a Avantor representative for more information on additional options.
Customers Who Bought This Also Bought