ContactUsLinkComponent
Pig Blue Lightweight Absorbent Mat Pads 50/Bx
Pig Blue® Absorbent Mat Pad in Dispenser Box, New Pig
Catalog #: 89132-766
Supplier:  New Pig
Pig Blue Lightweight Absorbent Mat Pads 50/Bx
Pig Blue® Absorbent Mat Pad in Dispenser Box, New Pig
Catalog #: 89132-766
Supplier:  New Pig
Specifications
  • Description:
    Lightweight, 50 Pads
  • Cat. no.:
    89132-766
  • Dimensions:
    38.1 x 48.3 cm (15 x 19")
  • Supplier No.:
    BLU107
Specifications
About this item
Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
Contains 75% recycled fibers; ideal in situations where green products are desired or required. Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08. Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Heavyweight Pig Blue construction is super absorbent for your most demanding tasks and large-volume cleanups. Lightweight Pig Blue construction is ideal for your everyday tasks and cleanups.
FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent. Stays together, even when picking up or shifting a fully saturated mat.
Customers Who Bought This Also Bought