Specifications
- Description:BTXpress Solution Only, 2 mL
- Size:2.0 ml
- No. of Reactions:20
- Supplier No.:45-0801
Specifications
About this item
Single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered 'hard to transfect' by chemical and other non-viral methods.
- High efficiency transfection of difficult cell lines
- High cell viability
- Low toxicity resulting in improved cell viability
- Single buffer for all cell types
- One buffer used in place of standard electroporation buffers for all mammalian cell types
BTXpress Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you.
As a universal solution, the BTXpress Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXpress Electroporation Solution is offered as a kit including the BTX Cuvettes Plus with transfer pipettes or as a buffer alone.