MicroGuard CE™ Coveralls, International ENVIROGUARD™
Catalog #: 89127-728
Supplier: International Enviroguard
Specifications
- Clothing Size:L
- Garment application:Cleanroom,Laboratory
- Color:White
- Description:Coverall with Attached Hood and Boots
- Cat. no.:89127-728
- Packaging:Clean Processed, Individually Packaged
- Supplier No.:CE8019CI-L
Specifications
About this item
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting
Coveralls are white with tunnelized wrists and ankles.
Tear-resistant. Recyclable.
Customers Who Bought This Also Bought