ViroGuard® Blue Coveralls with Hood and Boot, Elastic Wrist and Back, Front Zipper with Storm Flap, International Enviroguard™
Catalog #: 89127-662
Supplier: International Enviroguard
Specifications
- Clothing Size:M
- Garment application:Medical,Industrial,Clinical,Waste water,Laboratory,Pharmaceutical
- Closure type:Zip closure
- Color:Blue
- Description:ViroGuard® Coverall with Hood and Boot, Elastic Wrist and Back, Front Zipper with Storm Flap
- Cat. no.:89127-662
- Chest Size:36 - 39"
- Inseam:28"
- Supplier No.:2404-M
Specifications
About this item
Disposable Coverall with fabric that has been tested for protection against blood, blood-borne pathogens, and many other liquid hazards.
- Strong fabric provides excellent abrasion resistance and resists tears
- Helps to keep the wearer cool and avoid heat stress through enhanced moisture vapor transmission
- Attached hood and boots with elastic wrists for snug, secure fit
- Zipper front with sealable storm flap for an additional layer of protection
- Blue color makes stains and splashes on fabric less conspicuous
Fabric passes ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and blood-borne pathogens. Roomy, seamless shoulder area resists tears and provides more protection in key exposure area. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with user for more comfort and better tear resistance.
Recommended applications include emergency response, category 3 black water, tank cleaning, crime scene remediation, and animal research.
Customers Who Bought This Also Bought