Specifications
- Color:Natural
- Cat. no.:89125-442
- Growth Area:2.2 cm2 Per Compartment
- Working Volume:0.5 ml
- No. of Compartments:4
- Surface Treatment:Tissue culture (TC)
- Supplier No.:627870
- Total Volume:1.5 ml
Specifications
About this item
The CELLview™ cell culture dish is made from high-grade polystyrene combinded with an integrated glass bottom.
- Sterilized by irradiation
- Free of RNase, DNase, and human DNA
- Noncytotoxic
- Nonpyrogenic
The innovative design guarantees a single-plane, flat bottom with a consistent working distance and maximal planarity
CELLview™ combines the conveniance of a standard size 35mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures.
Glass bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN ISO 719) Glass thickness 175 um +/- 15um. Manufactured according to ISO 8255-1; 1986
The subdivided version of the CELLviewCell Culture Dish enables simultaneous multiplex analyzes of different cell lines, varioius stimulations, or diverse transfections.