Axygen® Automation Tips for Qiagen BioRobot® Series, Corning
Catalog #: 89124-962
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:300 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Tip Color:Black
- Sterile:-
- Supplier No.:QT-300-CBK-R
Specifications
About this item
Axygen® offers a comprehensive line of tips for Qiagen® automation systems.
- Black tips are conductive for liquid level sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Customers Who Bought This Also Bought