Axygen® Disposable Reagent Reservoirs, Corning
Catalog #: 89108-018
Supplier: Corning
Specifications
- Capacity:25 ml
- Sterility:Non sterile
- Description:Disposable Reagent Reservoir
- Environmentally Preferable:Low Manufacturing Impact
- Material:Plastic
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Packaging:100/case
- Supplier No.:RES-2CV-25
Specifications
About this item
These disposable reagent reservoirs are sterile and certified nonpyrogenic
They feature a sharp-V bottom that allows efficient extraction of the reagent. They also feature four pour-off corners and a stacking lug for easy separation
Customers Who Bought This Also Bought