VWR® Basic SPP Beard Covers
Catalog #: 89107-778
Supplier: VWR International
Specifications
- Clothing Size:One size
- Color:White
- Description:Latex-free
- Tensile strength:>30 (Elongation Percentage)
- Particle shedding (Helmke Drum):Level II
- Weight:10.0 g/m²
Specifications
About this item
VWR® Basic protection SPP beard covers are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable
- Economical first-line barrier
- Non-linting
Elastic band provides secure fit. Available in latex or latex-free formulation. Beard covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They protect against dirt, grime, and certain dry particulates in nonhazardous environments. These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms.
Customers Who Bought This Also Bought