ContactUsLinkComponent
VWR Weighted Tape Dispenser
VWR® Weighted Tape Dispenser
Catalog #: 89098-148
Supplier:  VWR International
VWR Weighted Tape Dispenser
VWR® Weighted Tape Dispenser
Catalog #: 89098-148
Supplier:  VWR International
Specifications
  • Description:
    Weighted tape dispenser
  • Cat. no.:
    89098-148
Specifications
About this item
Single-roll dispenser is designed with an adjustable core size to hold either a 12.7m (41.7') long roll of tape on a 25.4 mm (1") core or a 54.9m (180') long roll of tape on a 76.2 mm (3") core.
  • Adjustable core size
  • Heavy weighted base
  • Stable and secure support on any flat surface
The heavy weighted base provides increased stability and rests securely on any flat surface.
Ordering information: Tape and labels sold separately.
Customers Who Bought This Also Bought