Order Entry
ContactUsLinkComponent
VWR Red Border Tape Labels
VWR® Red Border Tape Labels
Catalog #: 89098-108
Supplier:  VWR International
VWR Red Border Tape Labels
VWR® Red Border Tape Labels
Catalog #: 89098-108
Supplier:  VWR International

Specifications

  • Description:
    Border tape labels
  • Cat. no.:
    89098-108
  • Dimensions:
    63.5×38.1 mm (2.5×1.5")

Specifications

About this item

Designed to aid in the identification of containers, shelves, and other standard laboratory or workplace items.

  • Self-adhesive tape design
  • Distinctive coloring for quick identification

Designed to aid in the identification of containers, shelves, and other standard laboratory or workplace items.

Labels are printed on a white. These labels can be written on with pen, pencil, or permanent marker. They can be easily removed and will not leave a sticky residue.

Ordering information: Supplied on individual rolls, each containing 12.7 m (41.7') of tape.
Customers Who Bought This Also Bought