Order Entry
ContactUsLinkComponent
VWR Tape
VWR® General Purpose Laboratory Labeling Tape
Catalog #: 89098-022
Supplier:  VWR International
VWR Tape
VWR® General Purpose Laboratory Labeling Tape
Catalog #: 89098-022
Supplier:  VWR International

Specifications

  • Length:
    54.9 m (180')
  • Width:
    19 mm (¾")
  • Color:
    Aqua

Specifications

About this item

VWR® General purpose laboratory labeling tape sticks to virtually any surface, can be marked with most writing utensils, and reliably withstands extreme temperature variations.

  • Vinyl-coated, soft satin-finish paper
  • Adhesive sticks to virtually any surface
  • Tape removal leaves no residue
  • Available in two roll sizes

A wide assortment of vinyl-coated, soft satin-finish paper tape that sticks to virtually any surface. It can be written on with pencil, fiber-tip or ball point pen, and lab markers. This durable, self-adhesive tape is ideal for general laboratory use, and is excellent for applications in refrigeration. It can withstand temperatures from -23...+121 °C (-10...+250 °F) and has an application temperature as low as 0 °C (32 °F). The adhesive backing will peel off easily and leaves no residue.

Ordering information: Available in two roll sizes. Rainbow packs contain blue, green, orange, pink, red, yellow, and white tape.
Customers Who Bought This Also Bought