VWR® Biohazard Labels
Catalog #: 89097-852
Supplier: VWR International
Specifications
- Description:Biohazard labels
- Cat. no.:89097-852
- Dimensions:76.2×50.8 mm (3×2")
Specifications
About this item
Labels aid in general workplace safety and efficiency by identifying contaminated equipment containers, rooms, or materials.
- Quick and easy identification
- Bold symbols declare biohazard warning from a distance
A biohazard symbol along with the term are printed in bold black atop a fluorescent red background.
Certifications: Labels are manufactured in accordance with various federal and state regulations, including OSHA safety standards.
Ordering information: Labels are available on rolls of 250 or 500 labels. One roll per case.
Customers Who Bought This Also Bought