KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
Catalog #: 89097-368
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Specifications
- Capacity:100 ml (3.4 oz.)
- Graduation Interval:10 ml
- Graduation range:20 – 80 mL
- Height:100 mm
- Cat. no.:89097-368
- Outer Diameter:56 mm
- Supplier No.:KC14395-100
Specifications
About this item
Ideal for general laboratory use including mixing, storing or transporting culture media, chemicals, or solvents.
- 30 mm Int.Ø. opening
- Blue polypropylene linerless GL 45 screw thread cap
- Enhanced graduations and marking spot made with chemically resistant white enamel paint
Autoclavable KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Blue caps and clear pour rings included.
Customers Who Bought This Also Bought