Order Entry
ContactUsLinkComponent
VWR* Static-Away* Brush
VWR® StaticAway™ Antistatic Brush
Catalog #: 89094-716
Supplier:  VWR International
VWR* Static-Away* Brush
VWR® StaticAway™ Antistatic Brush
Catalog #: 89094-716
Supplier:  VWR International

Specifications

  • Handle Material:
    Plastic
  • Description:
    Long-Handle Brush
  • Brush Head Width:
    2.5 cm (1")
  • Cat. no.:
    89094-716
  • Handle Length:
    12.7 cm (5")

Specifications

About this item

Brushes clean by dissipating static and whisking away harmful dust, lint, and dirt.

  • Provide accurate results by eliminating pan weight variances caused by static
  • Long-handled brushes allow access to difficult places

Ideal for cleaning balance pans, optics, cuvettes, slides, film, microscopes, electronics, and computer monitors. Bristles will not scratch critical surfaces. Brushes drain static and clean with synthetic conductive fibers and a conductive composite handle. The compact brush is suitable for electronic balances, providing accurate results by eliminating pan weight variances caused by static.

Customers Who Bought This Also Bought