Order Entry
ContactUsLinkComponent
PCR Mini-Cooler
PCR Mini Cooler, with Transparent Lid, BrandTech®
Catalog #: 89093-070
PCR Mini-Cooler
PCR Mini Cooler, with Transparent Lid, BrandTech®
Catalog #: 89093-070

Specifications

  • Cat. no.:
    89093-070
  • Supplier No.:
    781260

Specifications

About this item

The mini PCR cooler keeps samples at 4 ºC for approximately 3 hours.

  • Maintains 4 ˚C for master mix stability
  • Color indicates temperature
  • Holds 0.2 ml tubes, strips or plates
  • For use with PCR tubes 0.2 mL single, 8-strips, 12-strips and 96-well PCR plates

The insulating gel changes from violet to pink at 7 ºC. Suitable for 0.2 ml single tubes, 8-strips, and 12-strips, as well as 96-well PCR plates.

Customers Who Bought This Also Bought