Order Entry
ContactUsLinkComponent
CARDBOARD CRYOGENIC BOX CS15
WHEATON® CryoFile® and CryoFile®XL Storage Boxes, DWK Life Sciences
Catalog #: 89092-376
Small Business Enterprise
CARDBOARD CRYOGENIC BOX CS15
WHEATON® CryoFile® and CryoFile®XL Storage Boxes, DWK Life Sciences
Catalog #: 89092-376
Small Business Enterprise

Specifications

  • Dimensions:
    130×130×52 mm
  • Color:
    Green
  • Description:
    CryoFile® Storage Box
  • Cat. no.:
    89092-376
  • For Vials:
    1.2 and 2 ml vials
  • Supplier No.:
    W651600

Specifications

About this item

These CryoFile® and CryoFile®XL storage boxes provide high-quality storage in cryogenic temperatures.

  • Partitions numbered from 1 to 81 for easy content identification
  • Numbering system printed on lid and bottom of box
  • Six colors provide easy sample identification
  • Water repellent allows for longer durability
  • For use with vapor phase of liquid nitrogen

Directional holes in bottom allow for drainage and orientation of bottom to top of box.

Customers Who Bought This Also Bought