ContactUsLinkComponent
RETRIEVER MAGNET STIR BAR 1/CS
Corning® Magnetic Stir Bar Retriever, Corning
Catalog #: 89090-234
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
RETRIEVER MAGNET STIR BAR 1/CS
Corning® Magnetic Stir Bar Retriever, Corning
Catalog #: 89090-234
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Specifications
  • Description:
    Magnetic Stir Bar Retriever
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    89090-234
  • Supplier No.:
    6970SR
Specifications
About this item
The stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory.
Customers Who Bought This Also Bought