Corning® Pasteur Pipettes
Catalog #: 89090-042
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Length:146.05 mm (5³/₄")
- O.D.:6.5 mm
- Sterility:Non sterile
- Description:Disposable pasteur pipettes
- Material:Borosilicate glass
- Supplier No.:7095D-5X
Specifications
About this item
Soda-lime or borosilicate glass.
- Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
- Features a constriction one inch below the top to facilitate plugging
- Available in convenient bulk packs
These disposable glass Pasteur pipets are supplied in convenient bulk-packs. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.
Customers Who Bought This Also Bought