Corning® Clear Bottom, Black and White, Polystyrene Microplates, 384 Well, Corning
Catalog #: 89089-854
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Non sterile
- Well Volume:50 µl
- Color:Black with Clear Bottom
- Description:Corning® Low Volume 384 Well, Black with Clear Flat Bottom, Polystyrene, without Lid
- Environmentally Preferable:Low Manufacturing Impact
- No. of wells:384
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Packaging:10/Bag
- Well Shape:Flat-Bottom
- Surface Treatment:Untreated
- Supplier No.:3540
Specifications
About this item
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Customers Who Bought This Also Bought