Specimen Zipper Bags with Pouch and Biohazard Message, Associated Bag
Catalog #: 89085-396
Supplier: Associated Bag
Specifications
- W×L:6×9"
- Thickness:2 mil
- Cat. no.:89085-396
- Supplier No.:14-93
Specifications
About this item
Sure-seal zipper bag holds specimen and protects against accidental spills.
Separate document pouch keeps documents with specimens during transport and storage. Meets current labeling requirements of OSHA standard 29CFR1910.1030 regarding biohazard symbols.
Customers Who Bought This Also Bought