Order Entry
ContactUsLinkComponent
Hirschmann volumetric pipette
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Catalog #: 89085-234
Supplier:  Hirschmann
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Catalog #: 89085-234
Supplier:  Hirschmann

Specifications

  • Capacity:
    50 ml
  • Description:
    Serialized Pipets
  • Cat. no.:
    89085-234
  • Tolerance:
    ±0.05 ml
  • Band Color:
    White
  • Supplier No.:
    134G50S

Specifications

About this item

Pipettes are constructed of borosilicate glass and feature black enamel lettering and a graduation line.

Designed to deliver according to ASTM Specification E969, Class A requirements. Color-coded for ease in sorting and selection according to ASTM E1273. Serialized pipettes include a Certificate of Identification and Accuracy. All pipettes feature lot number and production year printed onto the pipette for QA purposes.

Customers Who Bought This Also Bought