Adapter, Outer Socket Joint, Ace Glass Incorporated
Catalog #: 89082-554
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Outer Socket Joint Adapter
- Cat. no.:89082-554
- SJ Joint No.:25/15
- Tube Size:10 mm
- Ace-thred no.:-
- Supplier No.:5221-05
Specifications
About this item
This series of adapters is designed for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
- Outer socket joints
- Various Ace-Thred and tube ends
- Fabricated from borosilicate glass
Adapters feature 28/15 and 35/25 sockets with various Ace-Thred and tube ends.
Customers Who Bought This Also Bought