Bath Oil, Ace Glass
Catalog #: 89078-216
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:
Specifications
- Color:Amber
- Description:Bath Oil, Hi-Temp
- Cat. no.:89078-216
- Volume:1.8L (606/7 oz.)
- Supplier No.:14115-14
Specifications
About this item
An extremely stable, medium viscosity silicone heating bath oil
Will not show discoloration after 40 hours at maximum temperature.
Ordering information: Available in two temperature ranges: Low Temp — maximum 180°C; or High Temp — maximum 230°C. Low temp oil color is CLEAR. High temp oil color is AMBER.
Customers Who Bought This Also Bought