Order Entry
ContactUsLinkComponent
13385 GAUGE, Pressure
Gauges, Pressure, Ace Glass
Catalog #: 89077-686
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
13385 GAUGE, Pressure
Gauges, Pressure, Ace Glass
Catalog #: 89077-686
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Gauge (only), Pressure, 1/4" Male NPT Bottom Fitting
  • Material:
    Brass, Internal
  • Size:
    2.5" dia.
  • Cat. no.:
    89077-686
  • Range:
    0 - 400 psi
  • Supplier No.:
    13385-12

Specifications

About this item

Pressure gauges for monitoring pressure in several pieces of apparatus such as pressure vessels, pressure reactors and chromatography columns

Pressure ranges vary from 0-60psi and up to 0-400psi. Gauges have different NPT end fittings and internal materials, as well as various dial ranges. Some come with nylon adapters to match to Ace-Threads, NPT threads and 1/4-UNF threads. One style comes complete with a three-way tubing connector for in-line connection. 89077-700 is a compound gauge for both pressure and vacuum applications.

Customers Who Bought This Also Bought