Order Entry
ContactUsLinkComponent
12770 TUBE FITTINGS, CONNECTORS, PTFE
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Catalog #: 89077-510
Supplier:  Ace Glass
Small Business Enterprise
12770 TUBE FITTINGS, CONNECTORS, PTFE
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
Catalog #: 89077-510
Supplier:  Ace Glass
Small Business Enterprise

Specifications

  • Connection type:
    Connector
  • Dimensions:
    18 W x 43.2L mm
  • Description:
    PTFE Connector, Fits Tubing O.D. 6.4 mm (1/4")
  • Cat. no.:
    89077-510
  • NPT:
    ¼"
  • Supplier No.:
    12770-24

Specifications

About this item

These all-PTFE fittings have a male NPT (MNPT) thread at one end and either tube compression, a hose barb, or MNPT thread at the other end.

Customers Who Bought This Also Bought