Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
Catalog #: 89077-390
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Female Coupling Tube to FNPT (with 6.4 mm Pipe Thread)
- Cat. no.:89077-390
- Fits Tubing O.D.:6.4 mm (¼")
- Supplier No.:12706-20
Specifications
About this item
These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.
Customers Who Bought This Also Bought